Learn More
Medchemexpress LLC Ethoxyquin dimer | 74681-77-9 | 98.70% | 432.60 | 1 MG
SDP
Catalog No. 5000349335
Encompass_Preferred
Ethoxyquin dimer is a dimer of Ethoxyquin and functions as an antioxidant. It can accumulate more readily in liver and adipose tissue compared to Ethoxyquin. Ethoxyquin at doses above 10 mg/kg has been shown to induce hepatotoxicity in mice.
  • Functions as an antioxidant
  • Accumulates more readily in liver and adipose tissue compared to Ethoxyquin
  • Intended for research use only
Catalog No. 50-003-49335 Supplier Medchemexpress LLC Supplier No. HY1382911MG
Ethoxyquin dimer is a dimer of Ethoxyquin and functions as an antioxidant. It can accumulate more readily in liver and adipose tissue compared to Ethoxyquin. Ethoxyquin at doses above 10 mg/kg has been shown to induce hepatotoxicity in mice.
  • Functions as an antioxidant
  • Accumulates more readily in liver and adipose tissue compared to Ethoxyquin
  • Intended for research use only