Learn More
Medchemexpress LLC HY-N4317 5mg Medchemexpress, Ethoxysanguinarine CAS:28342-31-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN43175MG
Encompass_Preferred
Medchemexpress, HY-N4317 5mg Ethoxysanguinarine CAS:28342-31-6 Ethoxysanguinarine is a benzophenanthridine alkaloid natural product that is mainly found in Macleaya cordata. Ethoxysanguinarine inhibits viability and induces apoptosis of colorectal cancer cells by inhibiting protein phosphatase 2A (CIP2A). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2934