Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethoxytriethylsilane, 97%

Catalog No. AAL1665614
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1665614 25 g
AAL1665606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1665614 Supplier Thermo Scientific Chemicals Supplier No. L1665614
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 597-67-1 | C8H20OSi | 160.332 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 597-67-1
Molecular Formula C8H20OSi
Molecular Weight (g/mol) 160.332
MDL Number MFCD00054718
InChI Key DFJDZTPFNSXNAX-UHFFFAOYSA-N
Synonym triethylethoxysilane, silane, ethoxytriethyl, ethoxy triethyl silane, acmc-1al7t
PubChem CID 284135
IUPAC Name ethoxy(triethyl)silane
SMILES CCO[Si](CC)(CC)CC

Specifications

Boiling Point 154°C to 155°C
Quantity 25 g
Refractive Index 1.414
Sensitivity Moisture sensitive
Density 0.816
UN Number UN1993
Beilstein 1735027
Formula Weight 160.33
Percent Purity 97%
Chemical Name or Material Ethoxytriethylsilane
Hazard Category H226-H315-H319-H335
Hazard Statement GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.