Learn More
Medchemexpress LLC HY-B1480 100mg Medchemexpress, Ethoxzolamide CAS:452-35-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB1480100MG
Encompass_Preferred
Medchemexpress, HY-B1480 100mg Ethoxzolamide CAS:452-35-7 Ethoxzolamide is a carbonic anhydrase inhibitor with K i of 1 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2949