Learn More
Medchemexpress LLC Ethyl 1-(2-bromoacetyl)cyclopropanecarboxylate | 129306-05-4 | 97.1% | 235.08 | 1 G
SDP
Catalog No. 5000404106
Encompass_Preferred
Ethyl 1-(2-bromoacetyl)cyclopropanecarboxylate is a drug intermediate for the synthesis of various active compounds.
  • Identified for laboratory chemical use
  • Suitable for the manufacture of substances
  • For research use only
  • Store pure form at -20°C for 3 years, or 4°C for 2 years
  • Store in solvent at -80°C for 6 months, or -20°C for 1 month
Catalog No. 50-004-04106 Supplier Medchemexpress LLC Supplier No. HY419431G
Add to Cart
Add to Cart
Ethyl 1-(2-bromoacetyl)cyclopropanecarboxylate is a drug intermediate for the synthesis of various active compounds.
  • Identified for laboratory chemical use
  • Suitable for the manufacture of substances
  • For research use only
  • Store pure form at -20°C for 3 years, or 4°C for 2 years
  • Store in solvent at -80°C for 6 months, or -20°C for 1 month