Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 1,3-Dimethylpyrazole-5-carboxylate 96.0+%, TCI America™
SDP

Catalog No. E10011G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E10011G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E10011G Supplier TCI America Supplier No. E10011G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5744-40-1
Molecular Formula C8H12N2O2
Molecular Weight (g/mol) 168.20
MDL Number MFCD00233452
InChI Key ZYSGPOXVDOROJU-UHFFFAOYSA-N
Synonym ethyl 1,3-dimethyl-1h-pyrazole-5-carboxylate, ethyl 1,3-dimethylpyrazole-5-carboxylate, 1,3-dimethyl-5-ethoxycarbonylpyrazole, 1,3-dimethylpyrazole-5-carboxylic acid ethyl ester, 2,5-dimethyl-2h-pyrazole-3-carboxylic acid ethyl ester, ethyl1,3-dimethyl-1h-pyrazole-5-carboxylate, ethyl 1,3-dimethyl-5-pyrazolecarboxylate, ethyl 2,5-dimethyl-2h-pyrazole-3-carboxylate, 1h-pyrazole-5-carboxylic acid, 1,3-dimethyl-, ethyl ester, 1,3-dimethyl-1h-pyrazole-5-carboxylic acid ethyl ester
PubChem CID 138576
IUPAC Name ethyl 1,3-dimethyl-1H-pyrazole-5-carboxylate
SMILES CCOC(=O)C1=CC(C)=NN1C

Specifications

Color Yellow
Boiling Point 90°C
Quantity 1 g
Formula Weight 168.20
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl 1,3-Dimethylpyrazole-5-carboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.