Learn More
Sigma Aldrich Ethyl 1-(4-methoxybenzyl)-3-methyl-1H-pyrazole-5-carboxylate
Catalog No. JRD11661G Shop All MilliporeSigma Sigma Organic Chemistry Products
Click to view available options
Quantity:
1 g
Catalog No. Quantity
JRD11661G 1 g
1 options
Catalog No. JRD11661G Supplier Sigma Aldrich Supplier No. JRD11661G
Only null left
Add to Cart
Add to Cart
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
  • Reactant for synthesis of:
  • Dpp-4 inhibitors
  • Jak2 inhibitors
  • Serotonin and noradrenaline reuptake inhibitors
  • Human tryptase inhibitors
  • Anti-thrombotic nicecotamidesreactive for oxidative c-arylation of free (nh)-heterocycles via direct (sp3) c-h bond functionalization
Specifications
MDL Number MFCD08445940
Quantity 1 g