Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 1-Benzyl-3-oxo-4-piperidinecarboxylate Hydrochloride 98.0+%, TCI America™
SDP

Catalog No. E08551G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E08551G 1 g
E08555G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E08551G Supplier TCI America Supplier No. E08551G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 52763-21-0
Molecular Formula C15H20ClNO3
Molecular Weight (g/mol) 297.78
MDL Number MFCD00012792
InChI Key UQOMEAWPKSISII-UHFFFAOYNA-N
Synonym ethyl 1-benzyl-3-oxopiperidine-4-carboxylate hydrochloride, ethyl n-benzyl-3-oxo-4-piperidine-carboxylate hydrochloride, ethyl 1-benzyl-3-oxo-4-piperidinecarboxylate hydrochloride, 1-benzyl-4-ethoxycarbonyl-3-piperidone hydrochloride, ethyl 1-benzyl-3-oxo-piperidine-4-carboxylate hcl, ethyl 1-benzyl-3-oxo-piperidine-4-carboxylate hydrochloride, 1-benzyl-3-oxopiperidine-4-carboxylic acid ethyl ester hydrochloride, ethyl n-benzyl-3-oxopiperidine-4-carboxylate hydrochloride, zlchem 420, pubchem19671
PubChem CID 2723880
IUPAC Name hydrogen ethyl 1-benzyl-3-oxopiperidine-4-carboxylate chloride
SMILES [H+].[Cl-].CCOC(=O)C1CCN(CC2=CC=CC=C2)CC1=O

Specifications

Melting Point 165°C
Color White-Yellow
Quantity 1 g
Formula Weight 297.78
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Ethyl 1-Benzyl-3-oxo-4-piperidinecarboxylate Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.