Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 1-Benzyl-4-oxo-3-piperidinecarboxylate Hydrochloride Hydrate 98.0+%, TCI America™
SDP

Catalog No. B203025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B203025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B203025G Supplier TCI America Supplier No. B203025G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1454-53-1
Molecular Formula C15H20ClNO3
Molecular Weight (g/mol) 297.78
MDL Number MFCD00012800
InChI Key YPFMNHZRNXPYBG-UHFFFAOYNA-N
Synonym ethyl 1-benzyl-4-oxopiperidine-3-carboxylate hydrochloride, 1-benzyl-3-carbethoxy-4-piperidone hydrochloride, ethyl 1-benzyl-4-oxo-3-piperidinecarboxylate hydrochloride, 1-benzyl-3-ethoxycarbonyl-4-piperidone hydrochloride, 1-benzyl-3-carbethoxy-4-piperidone hcl, 1-benzyl-3-carbethoxy-piperidone hcl, 1-benzyl-3-ethoxycarbonyl-4-piperidone hcl, 1-benzyl-3-carboethoxy-4-piperidone hydrochloride, 1-benzyl-3-ethoxycarbonyl-4-oxopiperidine hydrochloride
PubChem CID 102623
IUPAC Name ethyl 1-benzyl-4-oxopiperidine-3-carboxylate hydrochloride
SMILES Cl.CCOC(=O)C1CN(CC2=CC=CC=C2)CCC1=O

Specifications

Melting Point 175°C
Quantity 25 g
Formula Weight 297.78
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Ethyl 1-Benzyl-4-oxo-3-piperidinecarboxylate Hydrochloride Hydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.