Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 1-Boc-3-oxopiperidine-4-carboxylate, 97%, Thermo Scientific Chemicals

Catalog No. AAH6410203
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6410203 1 g
AAH6410206 5 g
AAH64102MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH6410203 Supplier Thermo Scientific Chemicals Supplier No. H6410203

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 71233-25-5
Molecular Formula C13H21NO5
Molecular Weight (g/mol) 271.313
MDL Number MFCD09878815
InChI Key WCTXJAXKORIYNA-UHFFFAOYSA-N
Synonym 1-tert-butyl 4-ethyl 3-oxopiperidine-1,4-dicarboxylate, ethyl 1-n-boc-3-oxopiperidine-4-carboxylate, ethyl n-boc-3-oxopiperidine-4-carboxylate, 3-oxo-piperidine-1,4-dicarboxylic acid 1-tert-butyl ester 4-ethyl ester, ethyl 1-n-boc-3-oxo-piperidine-4-carboxylate, ethyl 1-boc-3-oxopiperidine-4-carboxylate, ethyl n-tert-butoxycarbonyl-3-oxo-piperidine-4-carboxylate, 1-tert-butyl 4-ethyl 3-oxo-1,4-piperidinedicarboxylate, 1,4-piperidinedicarboxylic acid, 3-oxo-, 1-1,1-dimethylethyl 4-ethyl ester, ksc915i7p
PubChem CID 15852989
IUPAC Name 1-O-tert-butyl 4-O-ethyl 3-oxopiperidine-1,4-dicarboxylate
SMILES CCOC(=O)C1CCN(CC1=O)C(=O)OC(C)(C)C

Specifications

Quantity 1 g
Beilstein 421506
Formula Weight 271.31
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Ethyl 1-Boc-3-oxopiperidine-4-carboxylate
missing translation for 'healthHazard1' H315-H319-H335
missing translation for 'healthHazard3' P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.