Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ethyl [(1-phenyl-1H-pyrazolo[3,4-d]pyrimidin-4-yl)thio]acetate | 335223-43-3 | MFCD02572734 | 99.2% | 314.36 g/mol | C15H14N4O2S | 1 ML
SDP

Catalog No. 5000284269
Encompass_Preferred

F1874-108 is a small-molecule inhibitor of brassinosteroid biosynthesis and signal transduction supplied for research use. It is provided as a ready-to-use solution and includes safety and technical documentation.

  • Inhibits brassinosteroid biosynthesis and signal transduction.
  • Supplied as a 10 mM solution in DMSO, 1 mL volume.
  • Reported purity 99.22%.
  • Molecular formula C15H14N4O2S; molecular weight 314.36 g/mol.
  • Includes safety data sheet and data sheet for handling and experimental details.

Catalog No. 50-002-84269 Supplier Medchemexpress LLC Supplier No. HYW59084910MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

F1874-108 is a small-molecule inhibitor of brassinosteroid biosynthesis and signal transduction supplied for research use. It is provided as a ready-to-use solution and includes safety and technical documentation.

  • Inhibits brassinosteroid biosynthesis and signal transduction.
  • Supplied as a 10 mM solution in DMSO, 1 mL volume.
  • Reported purity 99.22%.
  • Molecular formula C15H14N4O2S; molecular weight 314.36 g/mol.
  • Includes safety data sheet and data sheet for handling and experimental details.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.