Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 1-Piperidinepropionate 97.0+%, TCI America™
SDP

Catalog No. P130525ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P130525ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P130525ML Supplier TCI America Supplier No. P130525ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 19653-33-9
Molecular Formula C10H19NO2
Molecular Weight (g/mol) 185.27
MDL Number MFCD00006514
InChI Key MPAWVDTVWPPKJQ-UHFFFAOYSA-N
Synonym 1-Piperidinepropionic Acid Ethyl Ester, 3-(Piperidino)propionic Acid Ethyl Ester, Ethyl 3-(Piperidino)propionate
PubChem CID 88183
IUPAC Name ethyl 3-(piperidin-1-yl)propanoate
SMILES CCOC(=O)CCN1CCCCC1

Specifications

Color Yellow
Boiling Point 106°C
Quantity 25 mL
Formula Weight 185.27
Physical Form Liquid
Chemical Name or Material Ethyl 1-Piperidinepropionate
EINECSNumber (5)-0772
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.