Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 1-tert-Butoxycarbonyl-4-oxo-3-piperidinecarboxylate 97.0+%, TCI America™
SDP

Catalog No. E09835G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E09835G 5 g
E09831G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E09835G Supplier TCI America Supplier No. E09835G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 98977-34-5
Molecular Formula C13H21NO5
Molecular Weight (g/mol) 271.313
MDL Number MFCD01862189
InChI Key ABBVAMUCDQETDO-UHFFFAOYSA-N
Synonym 1-tert-Butoxycarbonyl-4-oxo-3-piperidinecarboxylic Acid Ethyl Ester, Ethyl 1-Boc-4-oxo-3-piperidinecarboxylate, 1-Boc-4-oxo-3-piperidinecarboxylic Acid Ethyl Ester, 1-tert-Butoxycarbonyl-3-ethoxycarbonyl-4-piperidone, 1-Boc-3-ethoxycarbonyl-4-piperidone
PubChem CID 6420876
IUPAC Name 1-O-tert-butyl 3-O-ethyl 4-oxopiperidine-1,3-dicarboxylate
SMILES CCOC(=O)C1CN(CCC1=O)C(=O)OC(C)(C)C

Specifications

Melting Point 64°C
Color Yellow
Quantity 5 g
Formula Weight 271.31
Percent Purity ≥97.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material Ethyl 1-tert-Butoxycarbonyl-4-oxo-3-piperidinecarboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.