Learn More
Medchemexpress LLC Ethyl 10-bromodecanoate | 55099-31-5 | 97.1% | 279.21 | 5 G
SDP
Catalog No. 5000401086
Encompass_Preferred
Ethyl 10-bromodecanoate acts as both an anti-inflammatory and hemostatic agent. It belongs to the linolenic acid family, which also encompasses compounds such as diethyl succinate and ethyl palmitate. Additionally, this compound has demonstrated antibacterial efficacy.
  • Functions as an anti-inflammatory agent.
  • Functions as a hemostatic agent.
  • Exhibits antibacterial properties.
  • A member of the linolenic acid family.
Catalog No. 50-004-01086 Supplier Medchemexpress LLC Supplier No. HY320885G
Add to Cart
Add to Cart
Ethyl 10-bromodecanoate acts as both an anti-inflammatory and hemostatic agent. It belongs to the linolenic acid family, which also encompasses compounds such as diethyl succinate and ethyl palmitate. Additionally, this compound has demonstrated antibacterial efficacy.
  • Functions as an anti-inflammatory agent.
  • Functions as a hemostatic agent.
  • Exhibits antibacterial properties.
  • A member of the linolenic acid family.