Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ethyl 11-dodecenoate | 76063-06-4 | >99.0% | 226.36 g/mol | C14H26O2 | 10 MG
SDP

Catalog No. 5000289274
Encompass_Preferred

Ethyl 11-dodecenoate is the ethyl ester of 11-dodecenoic acid (CAS 76063-06-4), provided as a small-quantity research reagent for synthetic and analytical use. It has formula C14H26O2 and molecular weight 226.36 g/mol. Store cold under inert atmosphere to maintain stability.

  • Ethyl ester of 11-dodecenoic acid (C14H26O2).
  • Molecular weight 226.36 g/mol.
  • Available in small research pack sizes, including 10 mg.
  • Suitable for synthetic chemistry and analytical applications.
  • Store at -20°C under nitrogen; in solvent: -80°C for 6 months or -20°C for 1 month.

Catalog No. 50-002-89274 Supplier Medchemexpress LLC Supplier No. HY16608610MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Ethyl 11-dodecenoate is the ethyl ester of 11-dodecenoic acid (CAS 76063-06-4), provided as a small-quantity research reagent for synthetic and analytical use. It has formula C14H26O2 and molecular weight 226.36 g/mol. Store cold under inert atmosphere to maintain stability.

  • Ethyl ester of 11-dodecenoic acid (C14H26O2).
  • Molecular weight 226.36 g/mol.
  • Available in small research pack sizes, including 10 mg.
  • Suitable for synthetic chemistry and analytical applications.
  • Store at -20°C under nitrogen; in solvent: -80°C for 6 months or -20°C for 1 month.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.