Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 2-(1-Fmoc-3-indolyl)-4-methylthiazole-5-carboxylate, 97%, Thermo Scientific™

Catalog No. AAH5837006
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5837006 5 g
AAH5837003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5837006 Supplier Thermo Scientific Chemicals Supplier No. H5837006

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1858255-80-7
Molecular Formula C30H24N2O4S
Molecular Weight (g/mol) 508.592
MDL Number MFCD22988949
InChI Key RRACILVOHVVHHF-UHFFFAOYSA-N
Synonym 9h-fluoren-9-ylmethyl 3-5-ethoxycarbonyl-4-methyl-1,3-thiazol-2-yl indole-1-carboxylate, ethyl 2-1-fmoc-3-indolyl-4-methylthiazole-5-carboxylate, ethyl 2-1-9h-fluoren-9-yl methoxy carbonyl-1h-indol-3-yl-4-methylthiazole-5-carboxylate
PubChem CID 71741368
IUPAC Name ethyl 2-[1-(9H-fluoren-9-ylmethoxycarbonyl)indol-3-yl]-4-methyl-1,3-thiazole-5-carboxylate
SMILES CCOC(=O)C1=C(N=C(S1)C2=CN(C3=CC=CC=C32)C(=O)OCC4C5=CC=CC=C5C6=CC=CC=C46)C

Specifications

Quantity 5 g
Formula Weight 510.61
Percent Purity 97%
Chemical Name or Material Ethyl 2-(1-Fmoc-3-indolyl)-4-methylthiazole-5-carboxylate
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.