Learn More
Medchemexpress LLC Ethyl 2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-pentadecadeuteriooctanoate | 1219798-38-5 | 98.5% | 187.36 g/mol | C10H5D15O2 | 100 MG
SDP
Supplier:  Medchemexpress LLC HYW015305S100MG
Encompass_Preferred
Ethyl octanoate-d15 is a deuterium-labeled analogue of ethyl octanoate used as an isotope-labeled reference standard for mass spectrometry and analytical chemistry. It serves as a stable internal standard for quantitative analyses, method development, and accuracy verification in GC-MS and LC-MS workflows.
  • Deuterium-labeled internal standard for mass spectrometry and quantitative analysis.
  • High isotopic enrichment and purity (98.5%).
  • Molecular formula C10H5D15O2; molecular weight 187.36 g/mol.
  • Liquid appearance with density 0.867 g/cm3 for convenient handling.
  • Provided with datasheet, COA, and SDS for traceability and quality control.
  • Suitable for method development, calibration, and accuracy verification.
Catalog No. 50-003-03671