Learn More
Medchemexpress LLC Ethyl 2,3,4,6-tetra-O-acetyl-alpha-D-thioglucopyranoside | 41670-79-5 | MFCD00797608 | 97.0% | 392.4 g/mol | C16H24O9S | 5 G
SDP
Catalog No. 5000299491
Encompass_Preferred
Ethyl 2,3,4,6-tetra-O-acetyl-α-D-thioglucopyranoside (CAS 41670-79-5) is an acetylated thioglycoside supplied as a solid research reagent. It is commonly used in carbohydrate chemistry and glycoside synthesis as a protected thioglycosyl donor or intermediate for synthetic transformations.
  • Acetylated thioglycoside useful for glycosylation and synthetic applications.
  • Molecular formula C16H24O9S; molecular weight 392.4 g/mol.
  • Typical purity ~97% as supplied.
  • Solid powder form; commonly supplied in laboratory pack sizes from mg to g.
  • Store refrigerated and, when provided, under inert gas to preserve stability.
  • Supplied for research use only with SDS and handling information.
Catalog No. 50-002-99491 Supplier Medchemexpress LLC Supplier No. HYW0398485G
Ethyl 2,3,4,6-tetra-O-acetyl-α-D-thioglucopyranoside (CAS 41670-79-5) is an acetylated thioglycoside supplied as a solid research reagent. It is commonly used in carbohydrate chemistry and glycoside synthesis as a protected thioglycosyl donor or intermediate for synthetic transformations.
  • Acetylated thioglycoside useful for glycosylation and synthetic applications.
  • Molecular formula C16H24O9S; molecular weight 392.4 g/mol.
  • Typical purity ~97% as supplied.
  • Solid powder form; commonly supplied in laboratory pack sizes from mg to g.
  • Store refrigerated and, when provided, under inert gas to preserve stability.
  • Supplied for research use only with SDS and handling information.