Learn More
Medchemexpress LLC Ethyl 2,3,4,6-tetra-O-acetyl-α-D-thioglucopyranoside | 41670-79-5 | MFCD00797608 | >98.0% | 392.4 g/mol | C16H24O9S | 250 MG
SDP
Catalog No. 5000303061
Encompass_Preferred
Ethyl 2,3,4,6-tetra-O-acetyl-α-D-thioglucopyranoside (CAS 41670-79-5) is an acetylated thioglycoside used as a biochemical reagent and synthetic intermediate in carbohydrate chemistry. It is employed in glycosylation studies, derivatization, and analytical method development and is supplied as a characterized solid with known molecular identity.
  • Used as a synthetic intermediate for glycoside synthesis.
  • Suitable for glycosylation and derivatization reactions.
  • High purity suitable for analytical and preparative work.
  • Stable solid form with defined molecular identity.
  • Characterized by formula C16H24O9S and molecular weight 392.4 g/mol.
Catalog No. 50-003-03061 Supplier Medchemexpress LLC Supplier No. 250MGHYW039848
Ethyl 2,3,4,6-tetra-O-acetyl-α-D-thioglucopyranoside (CAS 41670-79-5) is an acetylated thioglycoside used as a biochemical reagent and synthetic intermediate in carbohydrate chemistry. It is employed in glycosylation studies, derivatization, and analytical method development and is supplied as a characterized solid with known molecular identity.
  • Used as a synthetic intermediate for glycoside synthesis.
  • Suitable for glycosylation and derivatization reactions.
  • High purity suitable for analytical and preparative work.
  • Stable solid form with defined molecular identity.
  • Characterized by formula C16H24O9S and molecular weight 392.4 g/mol.