Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Ethyl 2,3-butadienoate

Catalog No. 4949921G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
49-499-21G 1 g
49-499-25G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 49-499-21G Supplier Sigma Aldrich Supplier No. 4949921G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Ethyl 2,3-butadienoate is an α-allenic ester
  • The reaction of ethyl 2,3-butadienoate with n-tosylated imines in the presence of dabco (1,4-diazabicyclo[2.2.2]octane) or dmap (4-dimethylaminopyridine) forms azetidine derivatives or novel dihydropyridine derivatives respectively
  • The performance of bifunctional n-acyl aminophosphines to catalyze the asymmetric [3+2] cycloaddition of phenylidenemalononitrile with ethyl 2,3-butadienoate has been evaluated.ethyl 2,3-butadienoate may be used in the synthesis of dihydropyrans by reacting with acyclic enones
  • It may also be used to synthesize spiranic heterocycles by reacting with heterocyclic bis-arylidene ketones via phosphine-catalyzed [3+2] annulations

Specifications

CAS 14369-81-4
Density 0.966 g/mL (at 25°C (literature))
Boiling Point 80°C (60 mmHg)
Molecular Formula C6H8O2
Refractive Index n20/D 1.46 (literature)
Linear Formula H2C=C=CHCO2C2H5
MDL Number MFCD01863567
Quantity 1 g
Molecular Weight (g/mol) 112.13
Percent Purity 95%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.