Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ethyl 2-(4-((2-amino-4-oxothiazol-5(4H)-ylidene)methyl)-2-methoxyphenoxy)acetate | 939228-52-1 | 98.5% | 50 MG
SDP

Catalog No. 5000231660
Encompass_Preferred

Ethyl 2-(4-((2-amino-4-oxothiazol-5(4H)-ylidene)methyl)-2-methoxyphenoxy)acetate (CCI-007) is a small-molecule research compound with cytotoxic activity against infant leukemia cells harboring MLL rearrangements. It is supplied for laboratory research use only.

  • Active against infant leukemia with MLL rearrangements (IC50 2.5-6.2 μM in sensitive cells).
  • Molecular formula C15H16N2O5S, molecular weight 336.36 g/mol.
  • Typical purity 98.5% (reported by supplier).
  • Storage: solid at 4°C, protect from light; in solvent, -80°C up to 6 months or -20°C up to 1 month.
  • Supplied for research use only and not for human use.
  • Available in multiple milligram pack sizes, including 50 mg.

Catalog No. 50-002-31660 Supplier Medchemexpress LLC Supplier No. HY12269850MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ethyl 2-(4-((2-amino-4-oxothiazol-5(4H)-ylidene)methyl)-2-methoxyphenoxy)acetate (CCI-007) is a small-molecule research compound with cytotoxic activity against infant leukemia cells harboring MLL rearrangements. It is supplied for laboratory research use only.

  • Active against infant leukemia with MLL rearrangements (IC50 2.5-6.2 μM in sensitive cells).
  • Molecular formula C15H16N2O5S, molecular weight 336.36 g/mol.
  • Typical purity 98.5% (reported by supplier).
  • Storage: solid at 4°C, protect from light; in solvent, -80°C up to 6 months or -20°C up to 1 month.
  • Supplied for research use only and not for human use.
  • Available in multiple milligram pack sizes, including 50 mg.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.