Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ethyl-2-(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)cyclohex-3-enyl)acetate | 1166829-70-4 | >97.0% | 294.19 | C16H27BO4 | 500 MG
SDP

Catalog No. 5000210702
Encompass_Preferred

Bpin-Cyclohexene-C-COOEt is a boron-pinacol substituted cyclohexene ethyl acetate used as a PROTAC linker in medicinal chemistry and targeted protein degradation research. The compound is supplied for research use only and is reported to have high purity and defined molecular characteristics.

  • used as a PROtac linker in targeted protein degradation research.
  • molecular formula: C16H27BO4.
  • molecular weight: 294.19 g/mol.
  • high purity (>97.0%).
  • suitable for synthesis of SMARCA2/4 degrader scaffolds and related intermediates.

Catalog No. 50-002-10702 Supplier Medchemexpress LLC Supplier No. HYW063924500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Bpin-Cyclohexene-C-COOEt is a boron-pinacol substituted cyclohexene ethyl acetate used as a PROTAC linker in medicinal chemistry and targeted protein degradation research. The compound is supplied for research use only and is reported to have high purity and defined molecular characteristics.

  • used as a PROtac linker in targeted protein degradation research.
  • molecular formula: C16H27BO4.
  • molecular weight: 294.19 g/mol.
  • high purity (>97.0%).
  • suitable for synthesis of SMARCA2/4 degrader scaffolds and related intermediates.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.