Learn More
Medchemexpress LLC Ethyl-2-(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)cyclohex-3-enyl)acetate | 1166829-70-4 | MFCD18383324 | ≥97.0% | 294.19 g/mol | C16H27BO4 | 250 MG
SDP
Catalog No. 5000209414
Encompass_Preferred
Bpin-Cyclohexene-C-COOEt is a borylated cyclohexene ethyl ester used as a PROTAC linker and reagent in small-molecule synthesis. It has CAS number 1166829-70-4, molecular formula C16H27BO4, molecular weight 294.19 g/mol, and is supplied with ≥97.0% purity. It is commonly provided in small research pack sizes for laboratory synthesis.
  • Borylated cyclohexene ethyl ester useful as a PROTAC linker.
  • Documented for use in synthesis of SMARCA2/4 degrader compounds.
  • High purity suitable for synthetic chemistry (≥97.0%).
  • Available in small research pack sizes, including 250 MG.
  • Contains a boronate (Bpin) functional group compatible with cross-coupling reactions.
Catalog No. 50-002-09414 Supplier Medchemexpress LLC Supplier No. 250MGHYW063924
Bpin-Cyclohexene-C-COOEt is a borylated cyclohexene ethyl ester used as a PROTAC linker and reagent in small-molecule synthesis. It has CAS number 1166829-70-4, molecular formula C16H27BO4, molecular weight 294.19 g/mol, and is supplied with ≥97.0% purity. It is commonly provided in small research pack sizes for laboratory synthesis.
  • Borylated cyclohexene ethyl ester useful as a PROTAC linker.
  • Documented for use in synthesis of SMARCA2/4 degrader compounds.
  • High purity suitable for synthetic chemistry (≥97.0%).
  • Available in small research pack sizes, including 250 MG.
  • Contains a boronate (Bpin) functional group compatible with cross-coupling reactions.