Learn More
Sigma Aldrich Ethyl 2-(4-fluorophenyl)-4-methyl-1,3-thiazole-5-carboxylate
Shop All MilliporeSigma Sigma Organic Chemistry Products
Description
- 4-bromo-n,n-bis(trimethylsilyl)aniline is an n-protected aryl reagent used in the synthesis of:
- Hydrophilic conjugated porphyrin dimers for one-photon and two-photon photodynamic therapy
- Functionalized organometallic polymer, poly(ferrocenylsilane)
- Borylanilines such as 4-(dimesitylboryl)aniline and 4-(dimesitylboryl)-3,5-dimethylaniline
- Silicon-containing oligomeric poly(imido-amides) (pias)
Specifications
Specifications
| Molecular Formula | C13H12FNO2S |
| Linear Formula | C13H12FNO2S |
| MDL Number | MFCD08443675 |
| Quantity | 25 mL |
| Molecular Weight (g/mol) | 265.3 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.