Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Ethyl 2,5-Dibromothiazole-4-carboxylate

Catalog No. SYX003521G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SYX003521G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SYX003521G Supplier Sigma Aldrich Supplier No. SYX003521G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Alcl3 promoted friedel-crafts acylation of 4-tert-butylbenzoyl chloride with mesitylene was investigated.4-tert-butylbenzoyl chloride was used in the synthesis of 9,9α-spirobifluorene-bridged bipolar systems containing 1,3,4-oxadiazole-conjugated oligoaryl and triaryllamine moieties
  • It was also used in the preparation of 2-(4-tert-butylphenyl)-5-(4-methoxyphenyl)-1,3,4-oxadiazole

Specifications

CAS 208264-60-2
Molecular Formula C6H5Br2NO2S
Linear Formula C6H5Br2NO2S
MDL Number MFCD09265487
Quantity 100 g
Molecular Weight (g/mol) 314.98
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.