Learn More
Cayman Chemical ethyl 2,5-Dihydroxybenzoate, Purity: ≥98%, CAS: 3943-91-7, 1 g
SDP
Catalog No. 502982760
Encompass_Preferred
A dual modulator of bone cell differentiation; promotes osteoblast differentiation and inhibits RANKL-activated osteoclastogenesis
Catalog No. 50-298-2760 Supplier Cayman Chemical Supplier No. 210131 G
A dual modulator of bone cell differentiation; promotes osteoblast differentiation and inhibits RANKL-activated osteoclastogenesis