Learn More
Medchemexpress LLC Ethyl 2,5-dihydroxybenzoate | 3943-91-7 | MFCD02114094 | 100.0% | 182.17 g/mol | C9H10O4 | 250MG
SDP
Catalog No. 5000236256
Encompass_Preferred
Ethyl gentisate (Ethyl 2,5-dihydroxybenzoate) is a small-molecule research reagent used in biochemical and cellular studies. It is a white solid with molecular formula C9H10O4, molecular weight 182.17 g/mol, and CAS number 3943-91-7. A certificate of analysis reports high purity (99.97%), and the compound is supplied with batch-level documentation for analytical work.
  • High purity (99.97%) indicated by certificate of analysis.
  • Documented molecular formula and molecular weight for accurate calculations.
  • Provided as a solid suitable for typical laboratory handling.
  • Batch-level documentation available to support quality control.
  • Appropriate for biochemical and small-molecule research applications.
Catalog No. 50-002-36256 Supplier Medchemexpress LLC Supplier No. HY124478250MG
Add to Cart
Add to Cart
Ethyl gentisate (Ethyl 2,5-dihydroxybenzoate) is a small-molecule research reagent used in biochemical and cellular studies. It is a white solid with molecular formula C9H10O4, molecular weight 182.17 g/mol, and CAS number 3943-91-7. A certificate of analysis reports high purity (99.97%), and the compound is supplied with batch-level documentation for analytical work.
  • High purity (99.97%) indicated by certificate of analysis.
  • Documented molecular formula and molecular weight for accurate calculations.
  • Provided as a solid suitable for typical laboratory handling.
  • Batch-level documentation available to support quality control.
  • Appropriate for biochemical and small-molecule research applications.