Learn More
Medchemexpress LLC Ethyl 2-(5-methylsulfanylthiophene-2-carbonyl)-3,4-dihydro-1H-isoquinoline-3-carboxylate | 1254944-66-5 | MFCD18782736 | 99.9% | C18H19NO3S2 | 10MG
SDP
Catalog No. 5000258775
Encompass_Preferred
SR8278 is a small-molecule REV-ERBα antagonist used in research to modulate circadian rhythm and metabolic pathways. It is provided as a solid with high reported purity and characterized physicochemical data for use in both in vitro and in vivo studies.
  • High reported purity suitable for research use.
  • Soluble in DMSO at high concentration; documented in vivo vehicle protocols available.
  • Powder storage stability at typical laboratory temperatures for extended periods.
  • Applicable as a tool compound for studies of circadian biology, metabolism, and related disease models.
  • Available in small research-scale quantities and ready-to-use DMSO solutions for convenience.
Catalog No. 50-002-58775 Supplier Medchemexpress LLC Supplier No. HY1441510MG
SR8278 is a small-molecule REV-ERBα antagonist used in research to modulate circadian rhythm and metabolic pathways. It is provided as a solid with high reported purity and characterized physicochemical data for use in both in vitro and in vivo studies.
  • High reported purity suitable for research use.
  • Soluble in DMSO at high concentration; documented in vivo vehicle protocols available.
  • Powder storage stability at typical laboratory temperatures for extended periods.
  • Applicable as a tool compound for studies of circadian biology, metabolism, and related disease models.
  • Available in small research-scale quantities and ready-to-use DMSO solutions for convenience.