Learn More
Sigma Aldrich Ethyl 2-amino-4-(3-nitrophenyl)thiophene-3-carboxylate
Shop All MilliporeSigma Sigma Organic Chemistry Products
Description
- Tridodecylamine(tda) can be used in the following applications:
- Extraction of picolinic acid, citric acid, silver, mercury, selenium, zinc, cobalt, iron and succinic acid from aqueous solutions by using various organic solvents as diluents
- Recovery of remazol red 3bs (r3bs) dye from wastewater via emulsion liquid membrane (elm) process by using tda as the carrier agent
- Synthesis of gemini surfactant-based ionic liquids by reacting with dibromo alkanes
Specifications
Specifications
| Molecular Formula | C13H12N2O4S |
| Linear Formula | C13H12N2O4S |
| Quantity | 10 mL |
| MDL Number | MFCD02730458 |
| Molecular Weight (g/mol) | 292.31 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.