Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 2-amino-4,5-dimethylthiophene-3-carboxylate, 96%, Thermo Scientific™

Catalog No. AAH3312003
Change view
Click to view available options
Quantity:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3312003 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH3312003 Supplier Thermo Scientific Chemicals Supplier No. H3312003

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 4815-24-1
Molecular Formula C9H13NO2S
Quantity 1g
MDL Number MFCD00085051
Synonym ethyl 2-amino-4,5-dimethyl3-thenoate, ethyl 2-amino-4,5-dimethyl-3-thiophenecarboxylate, 2-amino-4,5-dimethylthiophene-3-carboxylic acid ethyl ester, 2-amino-4,5-dimethyl-thiophene-3-carboxylic acid ethyl ester, 3-thiophenecarboxylic acid, 2-amino-4,5-dimethyl-, ethyl ester, 2-amino-3-carbethoxy-4,5-dimethylthiophene, 2-amino-4,5-dimethyl-3-ethoxycarbonyl-thiophene, ethyl-2-amino-4,5-dimethylthiophene-3-carboxylate, ethyl2-amino-4,5-dimethylthiophene-3-carboxylate, ethyl 2-amino-4,5-dimethyl-thiophene-3-carboxylate
InChI Key JYSDXODDWAQWJR-UHFFFAOYSA-N
SMILES CCOC(=O)C1=C(SC(=C1C)C)N
IUPAC Name ethyl 2-amino-4,5-dimethylthiophene-3-carboxylate
Molecular Weight (g/mol) 199.268
PubChem CID 78536
Formula Weight 199.27
Percent Purity 96%
Chemical Name or Material Ethyl 2-amino-4,5-dimethylthiophene-3-carboxylate
Show More Show Less
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 225-387-5
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.