Learn More
Medchemexpress LLC Ethyl 2-amino-4-methyl-5-((3-nitrophenyl)carbamoyl)-thiophene-3-carboxylate | 303141-21-1 | 99.7% | 10 MG
SDP
Catalog No. 5000232037
Encompass_Preferred
DC-S239 is a selective inhibitor of histone methyltransferase SET7 with reported in vitro activity (IC50 = 4.59 μM); supplied as a high-purity solid for research use.
  • Selective SET7 inhibitor with IC50 = 4.59 μM.
  • Reported antiproliferative activity in MCF7 and HL60 cells.
  • High purity suitable for biochemical and cellular studies.
  • Soluble in DMSO (125 mg/mL) for in vitro experiments.
  • Stable as a powder at -20°C for long-term storage.
Catalog No. 50-002-32037 Supplier Medchemexpress LLC Supplier No. HY12109310MG
DC-S239 is a selective inhibitor of histone methyltransferase SET7 with reported in vitro activity (IC50 = 4.59 μM); supplied as a high-purity solid for research use.
  • Selective SET7 inhibitor with IC50 = 4.59 μM.
  • Reported antiproliferative activity in MCF7 and HL60 cells.
  • High purity suitable for biochemical and cellular studies.
  • Soluble in DMSO (125 mg/mL) for in vitro experiments.
  • Stable as a powder at -20°C for long-term storage.