Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ethyl 2-amino-4-methyl-5-((3-nitrophenyl)carbamoyl)-thiophene-3-carboxylate | 303141-21-1 | 99.7% | 5 MG
SDP

Catalog No. 5000232258
Encompass_Preferred

DC-S239 is a small-molecule SET7 (SETD7) histone methyltransferase inhibitor used in epigenetics and cancer cell proliferation research.

  • Selective SET7 inhibitor with reported IC50 = 4.59 μM.
  • Anticancer activity; inhibits proliferation of MCF7, HL60, and MV4-11 cells.
  • Molecular formula C15H15N3O5S; molecular weight 349.36 g/mol.
  • Purity 99.7% as supplied.
  • Solid, yellow to orange powder; store powder at -20°C for long term.
  • Available in small research packs including 5 MG.

Catalog No. 50-002-32258 Supplier Medchemexpress LLC Supplier No. HY1210935MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

DC-S239 is a small-molecule SET7 (SETD7) histone methyltransferase inhibitor used in epigenetics and cancer cell proliferation research.

  • Selective SET7 inhibitor with reported IC50 = 4.59 μM.
  • Anticancer activity; inhibits proliferation of MCF7, HL60, and MV4-11 cells.
  • Molecular formula C15H15N3O5S; molecular weight 349.36 g/mol.
  • Purity 99.7% as supplied.
  • Solid, yellow to orange powder; store powder at -20°C for long term.
  • Available in small research packs including 5 MG.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.