Learn More
Medchemexpress LLC Ethyl 2-amino-4-methyl-5-[(3-nitrophenyl)formamido]thiophene-3-carboxylate | 303141-21-1 | 99.7% | 50 MG
SDP
Catalog No. 5000233735
Encompass_Preferred
DC-S239 is a selective small-molecule inhibitor of the histone methyltransferase SET7 (SETD7) used for research applications. It is reported to inhibit SET7 with an IC50 of 4.59 μM and is supplied as a solid for laboratory use.
  • Selective SET7 inhibitor with reported IC50 of 4.59 μM.
  • High purity (99.7%).
  • Molecular weight 349.36 g/mol; formula C15H15N3O5S.
  • Solid, yellow-orange physical form suitable for handling and storage.
  • Available as a 50 mg research vial.
Catalog No. 50-002-33735 Supplier Medchemexpress LLC Supplier No. HY12109350MG
DC-S239 is a selective small-molecule inhibitor of the histone methyltransferase SET7 (SETD7) used for research applications. It is reported to inhibit SET7 with an IC50 of 4.59 μM and is supplied as a solid for laboratory use.
  • Selective SET7 inhibitor with reported IC50 of 4.59 μM.
  • High purity (99.7%).
  • Molecular weight 349.36 g/mol; formula C15H15N3O5S.
  • Solid, yellow-orange physical form suitable for handling and storage.
  • Available as a 50 mg research vial.