Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 2-Amino-4-phenylthiazole-5-carboxylate 97.0+%, TCI America™
SDP

Catalog No. E07695G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E07695G 5 g
E07691G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E07695G Supplier TCI America Supplier No. E07695G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 64399-23-1
Molecular Formula C12H12N2O2S
Molecular Weight (g/mol) 248.3
MDL Number MFCD00022452
InChI Key OZMXFXOHCUEEPD-UHFFFAOYSA-N
Synonym ethyl 2-amino-4-phenylthiazole-5-carboxylate, ethyl 2-amino-4-phenyl-5-thiazolecarboxylate, 5-thiazolecarboxylic acid, 2-amino-4-phenyl-, ethyl ester, 2-amino-4-phenylthiazole-5-carboxylic acid ethyl ester, acmc-209nlv, 2-amino-4-phenyl-5-carboethoxythiazole, ethyl 2-amino-4-phenylthiazol-5-carboxylate, ethyl2-amino-4-phenylthiazole-5-carboxylate, ethyl 2-amino-4-phenyl-thiazole-5-carboxylate, ethyl 2-amino-4-phenylthiazol-5-yl-carboxylate
PubChem CID 73239
IUPAC Name ethyl 2-amino-4-phenyl-1,3-thiazole-5-carboxylate
SMILES CCOC(=O)C1=C(N=C(S1)N)C2=CC=CC=C2

Specifications

Melting Point 169°C
Color Yellow
Quantity 5 g
Formula Weight 248.30
Percent Purity ≥97.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Ethyl 2-Amino-4-phenylthiazole-5-carboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.