Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl (2-Amino-4-thiazolyl)acetate 98.0+%, TCI America™
SDP

Catalog No. A095525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A095525G 25 g
A0955500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A095525G Supplier TCI America Supplier No. A095525G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 53266-94-7
Molecular Formula C7H10N2O2S
Molecular Weight (g/mol) 186.23
MDL Number MFCD00005330
InChI Key SHQNGLYXRFCPGZ-UHFFFAOYSA-N
Synonym ethyl 2-amino-4-thiazoleacetate, ethyl 2-2-aminothiazol-4-yl acetate, ethyl 2-2-amino-1,3-thiazol-4-yl acetate, ethyl 2-aminothiazole-4-acetate, ethyl 2-amino-1,3-thiazol-4-yl acetate, ethyl 2-aminothiazol-4-acetate, 2-amino-4-thiazolyl acetic acid ethyl ester, 4-thiazoleacetic acid, 2-amino-, ethyl ester, ethyl 2-amino-4-thiazolyl acetate
PubChem CID 104454
IUPAC Name ethyl 2-(2-amino-1,3-thiazol-4-yl)acetate
SMILES CCOC(=O)CC1=CSC(N)=N1

Specifications

Melting Point 94°C
Quantity 25 g
Formula Weight 186.23
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Ethyl (2-Amino-4-thiazolyl)acetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.