Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sigma Aldrich Ethyl 2-amino-5-ethylthiophene-3-carboxylate
Shop All MilliporeSigma Sigma Organic Chemistry Products
Description
- 1,9-decadiene acts as comonomer and undergoes acyclic diene metathesis (admet) copolymerization with 1, 5-hexadiene to form random linear polybutadiene -polyoctenamer copolymers
- It undergoes admet copolymerization with divinyltetraethoxydisiloxane to yield siloxylene-vinylene-alkenylene copolymer.1,9-decadiene was used in synthesis and characterization of new telechelic polyoctenamers prepared by admet polymerization using grubbs catalyst
Specifications
Specifications
| CAS | 4507-13-5 |
| Molecular Formula | C9H13NO2S |
| Linear Formula | C9H13NO2S |
| MDL Number | MFCD00015559 |
| Quantity | 5 g |
| Molecular Weight (g/mol) | 199.27 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?