Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 2-Amino-5-methylthiophene-3-carboxylate 98.0+%, TCI America™
SDP

Catalog No. E11041G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E11041G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E11041G Supplier TCI America Supplier No. E11041G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

CAS 4815-32-1
Melting Point 49°C
Color Yellow
Molecular Formula C8H11NO2S
Quantity 1 g
MDL Number MFCD01922142
Synonym 2-amino-5-methyl-thiophene-3-carboxylic acid ethyl ester, ethyl 2-amino-5-methyl-3-thiophenecarboxylate, 2-amino-5-methylthiophene-3-carboxylic acid ethyl ester, 3-thiophenecarboxylic acid, 2-amino-5-methyl-, ethyl ester, ethyl2-amino-5-methyl-3-thiophenecarboxylate, 2-amino-5-methyl-3-thiophenecarboxylic acid ethyl ester, ethyl 2-amino-5-methyl-thiophene-3-carboxylate, ksc590m8t, aronis25101, 2-amino-5-methyl-thiophene-3-carboxylicacidethylester
InChI Key AYBUNZGJQVYGTM-UHFFFAOYSA-N
SMILES CCOC(=O)C1=C(SC(=C1)C)N
IUPAC Name ethyl 2-amino-5-methylthiophene-3-carboxylate
Molecular Weight (g/mol) 185.241
PubChem CID 4062885
Formula Weight 185.24
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Ethyl 2-Amino-5-methylthiophene-3-carboxylate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.