Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 2-aminocyclopenta[b]thiophene-3-carboxylate 99+%, Thermo Scientific™

Catalog No. AC292850010
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC292850010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC292850010 Supplier Thermo Scientific Chemicals Supplier No. 292850010

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4815-29-6
Molecular Formula C10H13NO2S
Molecular Weight (g/mol) 211.28
MDL Number MFCD00102063
InChI Key BOJXCJDYZJSPMZ-UHFFFAOYSA-N
Synonym ethyl 2-amino-5,6-dihydro-4h-cyclopenta b thiophene-3-carboxylate, 2-amino-5,6-dihydro-4h-cyclopenta b thiophene-3-carboxylic acid ethyl ester, ethyl 2-amino-4h,5h,6h-cyclopenta b thiophene-3-carboxylate, ethyl 2-aminocyclopenta b thiophene-3-carboxylate, ethyl 2-amino-5,6-dihydro-4h-cyclopenta b-thiophene-3-carboxylate, 4h-cyclopenta b thiophene-3-carboxylic acid, 2-amino-5,6-dihydro-, ethyl ester, ethyl 2-amino-4,5,6-trihydrocyclopenta 1,2-b thiophene-3-carboxylate, enamine_004067, ethyl 2-aminocyclopentathiophene-3-carboxylate
PubChem CID 264105
IUPAC Name ethyl 2-amino-4H,5H,6H-cyclopenta[b]thiophene-3-carboxylate
SMILES CCOC(=O)C1=C(N)SC2=C1CCC2

Specifications

Melting Point 90°C to 94°C
Infrared Spectrum Authentic
Quantity 1 g
Beilstein 18, V,12, 219
Formula Weight 211.28
Percent Purity ≥99.0%
Physical Form Crystalline Powder
Chemical Name or Material Ethyl 2-aminocyclopenta[b]thiophene-3-carboxylate, 99+%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.