Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 2-Amylacetoacetate 95.0+%, TCI America™
SDP

Catalog No. A04445ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A04445ML 5 mL
A044425ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A04445ML Supplier TCI America Supplier No. A04445ML

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 24317-94-0
Molecular Formula C11H20O3
Molecular Weight (g/mol) 200.278
MDL Number MFCD00059380
InChI Key XIGZBCUFFUBWDM-UHFFFAOYSA-N
Synonym 2-Acetylheptanoic Acid Ethyl Ester, 2-Amylacetoacetic Acid Ethyl Ester, Ethyl 2-Acetylheptanoate
PubChem CID 91162
IUPAC Name ethyl 2-acetylheptanoate
SMILES CCCCCC(C(=O)C)C(=O)OCC

Specifications

Color Yellow
Boiling Point 117°C
Quantity 5 mL
Formula Weight 200.28
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl 2-Amylacetoacetate
EINECSNumber (2)-1505
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.