Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 2-bromobutyrate, 97%, Thermo Scientific™

Catalog No. AAA1272930
Change view
Click to view available options
Quantity:
250g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1272930 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA1272930 Supplier Thermo Scientific Chemicals Supplier No. A1272930

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Ethyl 2-bromobutyrate, is an important raw material and an intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff. It is also used for agrochemical and pharmaceutical intermediate.

Solubility
Insoluble in water. Miscible with alcohol and ether

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Recommended storage temperature: 2 - 8°C. Light sensitive. Keep away from strong oxidizing agents, acids, bases, oxidizing agents, reducing agents.
TRUSTED_SUSTAINABILITY

Specifications

CAS 533-68-6
Density 1.324
Boiling Point 176°C to 178°C
Flash Point 58°C (136°F)
Molecular Formula C6H11BrO2
Refractive Index 1.447
Quantity 250g
MDL Number MFCD00000155
UN Number UN3272
Beilstein 969841
Sensitivity Light sensitive
Merck Index 14,1413
Synonym ethyl 2-bromobutyrate, dl-ethyl 2-bromobutyrate, 2-bromobutanoic acid ethyl ester, butanoic acid, 2-bromo-, ethyl ester, 2-bromobutyric acid ethyl ester, ethyl .alpha.-bromobutyrate, 2-bromo-butyric acid ethyl ester, butyric acid, 2-bromo-, ethyl ester, 2-bromo-butanoicaciethylester, ethyl alpha-bromobutyrate
Solubility Information Insoluble in water. Miscible with alcohol and ether
InChI Key XIMFCGSNSKXPBO-UHFFFAOYNA-N
SMILES CCOC(=O)C(Br)CC
IUPAC Name ethyl 2-bromobutanoate
Molecular Weight (g/mol) 195.06
PubChem CID 79039
Formula Weight 195.06
Percent Purity 97%
Chemical Name or Material Ethyl 2-bromobutyrate
Show More Show Less
Hazard Category H226-H314
Hazard Statement GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P240-P241-P242-P243-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
EINECSNumber 208-574-6
TSCA Yes
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.