Learn More
Accela Chembio Inc Ethyl 2-bromothiazole-4-carboxylate | 5g | 100367-77-9 | MFCD03788564 | 96% | Shelf Life: 1260 Days | Light Sensitive/moisture Sensitive/nitrogen Or Argon/+4
Catalog No. 50250498
Encompass_Preferred
Ethyl 2-bromothiazole-4-carboxylate | 5g | 100367-77-9 | MFCD03788564 | 96% | Shelf Life: 1260 Days | Light Sensitive/moisture Sensitive/nitrogen Or Argon/+4
Catalog No. 50-250-498 Supplier Accela Chembio Inc Supplier No. SY0044985G
Ethyl 2-bromothiazole-4-carboxylate | 5g | 100367-77-9 | MFCD03788564 | 96% | Shelf Life: 1260 Days | Light Sensitive/moisture Sensitive/nitrogen Or Argon/+4