Learn More
Medchemexpress LLC Ethyl 2-chloroacetoacetate | 609-15-4 | MFCD00009141 | 99.6% | 164.59 g/mol | C6H9ClO3 | 1kg
SDP
Catalog No. 5000269691
Encompass_Preferred
Ethyl 2-chloroacetoacetate (A 21960) is a Tn3 resolvase inhibitor and inhibit synapse formation between resolvase and two directly repeated res sites[1]
Catalog No. 50-002-69691 Supplier Medchemexpress LLC Supplier No. HYY03641KG
Ethyl 2-chloroacetoacetate (A 21960) is a Tn3 resolvase inhibitor and inhibit synapse formation between resolvase and two directly repeated res sites[1]