Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 2-chloroacetoacetate 97%, Thermo Scientific™

Catalog No. AC118232500
Change view
Click to view available options
Quantity:
250 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC118232500 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC118232500 Supplier Thermo Scientific Chemicals Supplier No. 118232500

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 609-15-4
Molecular Formula C6H9ClO3
Molecular Weight (g/mol) 164.59
MDL Number MFCD00009141
InChI Key RDULEYWUGKOCMR-UHFFFAOYNA-N
Synonym ethyl 2-chloroacetoacetate, 2-chloroacetoacetic acid ethyl ester, butanoic acid, 2-chloro-3-oxo-, ethyl ester, ethyl alpha-chloroacetoacetate, ethyl-2-chloracetoacetate, 2-chloro-3-oxobutyric acid ethyl ester, ethyl2-chloroacetoacetate, 2-chloro-3-oxobutanoic acid, ethyl ester, ethyl2-chloro-3-oxobutanoate, ethyl 2-chloro-3-oxobutyrate
PubChem CID 11858
IUPAC Name ethyl 2-chloro-3-oxobutanoate
SMILES CCOC(=O)C(Cl)C(C)=O

Specifications

Color Yellow
Density 1.19
Infrared Spectrum Authentic
Refractive Index 1.4410 to 1.4440 (20°C, 589nm)
Linear Formula 210-180-4NYCH3COCH(Cl)CO2C2H5
Quantity 250 mL
Beilstein 03, 662
Formula Weight 164.59
Percent Purity ≥96.0%
Physical Form Liquid
Chemical Name or Material Ethyl 2-chloroacetoacetate, 97%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.