Learn More
Medchemexpress LLC Ethyl 2-chloronicotinate | 1452-94-4 | MFCD00674129 | 100.0% | 185.61 g/mol | C8H8ClNO2 | 100 G
SDP
Catalog No. 5000302179
Encompass_Preferred
Ethyl 2-chloronicotinate is an organic biochemical reagent used as an intermediate in medicinal chemistry and agrochemical synthesis. Supplied as a high-purity liquid, it is intended for research and analytical applications where defined composition and reproducible performance are required.
  • High purity suitable for analytical use.
  • Useful intermediate for pharmaceutical and agrochemical synthesis.
  • Available in multiple pack sizes for small- and large-scale work.
  • Colorless to light yellow liquid with reported density 1.2445 g/cm3.
  • Molecular weight 185.61 g/mol and formula C8H8ClNO2.
Catalog No. 50-003-02179 Supplier Medchemexpress LLC Supplier No. HYW016216100G
Ethyl 2-chloronicotinate is an organic biochemical reagent used as an intermediate in medicinal chemistry and agrochemical synthesis. Supplied as a high-purity liquid, it is intended for research and analytical applications where defined composition and reproducible performance are required.
  • High purity suitable for analytical use.
  • Useful intermediate for pharmaceutical and agrochemical synthesis.
  • Available in multiple pack sizes for small- and large-scale work.
  • Colorless to light yellow liquid with reported density 1.2445 g/cm3.
  • Molecular weight 185.61 g/mol and formula C8H8ClNO2.