Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ethyl 2-chloronicotinate | 1452-94-4 | MFCD00674129 | 100.0% | 185.61 g/mol | C8H8ClNO2 | 25 G
SDP

Catalog No. 5000304147
Encompass_Preferred

Ethyl 2-chloronicotinate (CAS 1452-94-4) is an ethyl ester of 2-chloronicotinic acid used as a research-grade biochemical reagent and synthetic intermediate in medicinal chemistry and organic synthesis. Supplied as a high-purity liquid, it is suitable for analytical applications and small-scale synthesis.

  • High purity (99.96%) suitable for analytical work.
  • Molecular weight 185.61 g/mol.
  • Chemical formula C8H8ClNO2.
  • Colorless to light yellow liquid with density 1.2445 g/cm3.
  • Stable when stored as recommended: pure compound -20°C (3 years), 4°C (2 years).
  • Available in multiple small-scale package sizes for research use.

Catalog No. 50-003-04147 Supplier Medchemexpress LLC Supplier No. 25GHYW016216
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ethyl 2-chloronicotinate (CAS 1452-94-4) is an ethyl ester of 2-chloronicotinic acid used as a research-grade biochemical reagent and synthetic intermediate in medicinal chemistry and organic synthesis. Supplied as a high-purity liquid, it is suitable for analytical applications and small-scale synthesis.

  • High purity (99.96%) suitable for analytical work.
  • Molecular weight 185.61 g/mol.
  • Chemical formula C8H8ClNO2.
  • Colorless to light yellow liquid with density 1.2445 g/cm3.
  • Stable when stored as recommended: pure compound -20°C (3 years), 4°C (2 years).
  • Available in multiple small-scale package sizes for research use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.