Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ethyl 2-chloronicotinate | 1452-94-4 | MFCD00674129 | 100.0% | 185.61 g/mol | C8H8ClNO2 | 250 G
SDP

Catalog No. 5000303597
Encompass_Preferred

Ethyl 2-chloronicotinate (CAS 1452-94-4) is an organic intermediate and biochemical reagent used in medicinal chemistry and small-molecule synthesis. It functions as a chlorinated nicotinate ester for constructing heterocyclic scaffolds and performing ester-related transformations in research applications. Supplied for research use only.

  • High purity (99.96%) suitable for synthesis.
  • Molecular weight 185.61 g/mol.
  • Molecular formula C8H8ClNO2.
  • Available in multiple pack sizes, including 250 g.
  • Recommended storage: pure form -20°C for up to 3 years.

Catalog No. 50-003-03597 Supplier Medchemexpress LLC Supplier No. 250GHYW016216
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ethyl 2-chloronicotinate (CAS 1452-94-4) is an organic intermediate and biochemical reagent used in medicinal chemistry and small-molecule synthesis. It functions as a chlorinated nicotinate ester for constructing heterocyclic scaffolds and performing ester-related transformations in research applications. Supplied for research use only.

  • High purity (99.96%) suitable for synthesis.
  • Molecular weight 185.61 g/mol.
  • Molecular formula C8H8ClNO2.
  • Available in multiple pack sizes, including 250 g.
  • Recommended storage: pure form -20°C for up to 3 years.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.