Learn More
Medchemexpress LLC Ethyl 2-chloronicotinate | 1452-94-4 | MFCD00674129 | 100.0% | 185.61 g/mol | C8H8ClNO2 | 50 G
SDP
Catalog No. 5000302670
Encompass_Preferred
Ethyl 2-chloronicotinate (CAS 1452-94-4) is the ethyl ester of 2-chloronicotinic acid supplied as a high-purity biochemical reagent for organic synthesis, analytical standards, and life-science research. It is commonly used as a synthetic intermediate and analytical reference in medicinal chemistry and related research applications.
  • High purity suitable for analytical and synthetic applications.
  • Functions as a versatile intermediate in pharmaceutical and agrochemical synthesis.
  • Soluble in common organic solvents, facilitating synthetic reactions.
  • Offered in multiple pack sizes to support small-scale and bulk use.
  • Supplied with specification data to support quality control.
Catalog No. 50-003-02670 Supplier Medchemexpress LLC Supplier No. HYW01621650G
Ethyl 2-chloronicotinate (CAS 1452-94-4) is the ethyl ester of 2-chloronicotinic acid supplied as a high-purity biochemical reagent for organic synthesis, analytical standards, and life-science research. It is commonly used as a synthetic intermediate and analytical reference in medicinal chemistry and related research applications.
  • High purity suitable for analytical and synthetic applications.
  • Functions as a versatile intermediate in pharmaceutical and agrochemical synthesis.
  • Soluble in common organic solvents, facilitating synthetic reactions.
  • Offered in multiple pack sizes to support small-scale and bulk use.
  • Supplied with specification data to support quality control.