Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ethyl 2-chloropyrimidine-5-carboxylate | 89793-12-4 | 186.60 | 10 G
SDP

Catalog No. 5000445669
Encompass_Preferred

Ethyl 2-chloropyrimidine-5-carboxylate is a chemical compound commonly used for synthesis in laboratory research. This white to yellow solid is provided for research purposes only.

  • Molecular formula: C7H7ClN2O2
  • Molecular weight: 186.60
  • CAS number: 89793-12-4
  • Purity: 99.7%
  • Identified uses: Laboratory chemicals, manufacture of substances
  • Appearance: Solid, white to yellow
  • Melting/freezing point: 52-60°C
  • Boiling point: 314.2°C at 760 mmHg
  • Stable under recommended storage conditions
  • Storage: 4°C, sealed storage, away from moisture; in solvent: -80°C, 6 months; -20°C, 1 month

Catalog No. 50-004-45669 Supplier Medchemexpress LLC Supplier No. HYW00399510G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ethyl 2-chloropyrimidine-5-carboxylate is a chemical compound commonly used for synthesis in laboratory research. This white to yellow solid is provided for research purposes only.

  • Molecular formula: C7H7ClN2O2
  • Molecular weight: 186.60
  • CAS number: 89793-12-4
  • Purity: 99.7%
  • Identified uses: Laboratory chemicals, manufacture of substances
  • Appearance: Solid, white to yellow
  • Melting/freezing point: 52-60°C
  • Boiling point: 314.2°C at 760 mmHg
  • Stable under recommended storage conditions
  • Storage: 4°C, sealed storage, away from moisture; in solvent: -80°C, 6 months; -20°C, 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.