Learn More
Medchemexpress LLC Ethyl 2-cyano-2-(hydroxyimino)acetate | 3849-21-6 | 99.89% | 142.11 | 1 ML
SDP
Catalog No. 5000404552
Encompass_Preferred
Ethyl 2-cyano-2-(hydroxyimino)acetate (Oxyma) is a peptide coupling additive that suppresses racemization during peptide coupling and improves the coupling efficiency of both solution-phase and solid-phase peptide synthesis. It is for research use only.
  • Suppresses racemization during peptide coupling
  • Improves coupling efficiency for solution-phase synthesis
  • Improves coupling efficiency for solid-phase synthesis
Catalog No. 50-004-04552 Supplier Medchemexpress LLC Supplier No. HY4145610MM/1ML
Ethyl 2-cyano-2-(hydroxyimino)acetate (Oxyma) is a peptide coupling additive that suppresses racemization during peptide coupling and improves the coupling efficiency of both solution-phase and solid-phase peptide synthesis. It is for research use only.
  • Suppresses racemization during peptide coupling
  • Improves coupling efficiency for solution-phase synthesis
  • Improves coupling efficiency for solid-phase synthesis