Learn More
Medchemexpress LLC Ethyl 2-cyano-2-(hydroxyimino)acetate | 3849-21-6 | MFCD00002112 | 99.9% | 142.11 | 50 G
SDP
Supplier:  Medchemexpress LLC HY4145650G
Encompass_Preferred
Ethyl 2-cyano-2-(hydroxyimino)acetate (Oxyma) is a peptide coupling additive that suppresses racemization during peptide coupling and improves the coupling efficiency in both solution-phase and solid-phase peptide synthesis.
  • Suppresses racemization during peptide coupling.
  • Improves coupling efficiency of solution-phase peptide synthesis.
  • Improves coupling efficiency of solid-phase peptide synthesis.
  • Facilitates efficient solution-phase synthesis of Leu-enkephalin.
Catalog No. 50-004-03657